Dithiothreitol (DTT, Cleland's reagent)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3483-12-3
:27565-41-9
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00004877
:154.25 g/mol
Melting Pt:40…44 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together