
L(+)-Aspartic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:56-84-8
:
Formula:C₄H₇NO₄
MDL Number:MFCD00002616
:133.1 g/mol
Melting Pt:>300 °C
Density:1,66 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together