Order Entry
Northern Ireland
ContactUsLinkComponent
2-(Perfluoro-5-methylhexyl)ethyl methacrylate 97%
Irritant
2-(Perfluoro-5-methylhexyl)ethyl methacrylate 97%
Catalog # APOSPC6121M-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(Perfluoro-5-methylhexyl)ethyl methacrylate 97%
Catalog # APOSPC6121M-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :482.185 g/mol
Density:1,567 g/cm³ (25 °C)
 :GHS07
Storage Temperature:Refrigerator
CAS Number:50836-66-3
Formula:C₁₃H₉F₁₅O₂
MDL Number:MFCD00236098