Order Entry
Northern Ireland
ContactUsLinkComponent
cis-2-(Trifluoromethyl)cyclopropane-1-carboxylic acid 93%
Corrosive
Irritant
cis-2-(Trifluoromethyl)cyclopropane-1-carboxylic acid 93%
Catalog # APOSPC405624-100MG
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
cis-2-(Trifluoromethyl)cyclopropane-1-carboxylic acid 93%
Catalog # APOSPC405624-100MG
Supplier:  Apollo Scientific
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :154.087 g/mol
Storage Temperature:Refrigerator
UN:3265
ADR:8,II
 :GHS05
 :GHS07
CAS Number:1258652-18-4
Formula:C₅H₅F₃O₂
MDL Number:MFCD17977173