4-Chloro-3-fluoro-5-(trifluoromethyl)phenylacetonitrile 97%
Catalog # APOSPC302743-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:237.581 g/mol
Storage Temperature:Ambient
CAS Number:1431329-62-2
Formula:C₉H₄ClF₄N
MDL Number:MFCD23795335