3,6,6-Trimethyl-1,5,6,7-tetrahydro-4h-indazol-4-one 95%
Catalog # APOSOR929611-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:178.231 g/mol
Storage Temperature:Ambient
CAS Number:16315-16-5
Formula:C₁₀H₁₄N₂O
MDL Number:MFCD00474387