4-[(Propylcarbamoyl)amino]benzeneboronic acid, pinacol ester 98%
Catalog # APOSOR5972-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:304.192 g/mol
Storage Temperature:Ambient
CAS Number:874291-01-7
Formula:C₁₆H₂₅BN₂O₃
MDL Number:MFCD09027288