Order Entry
Northern Ireland
ContactUsLinkComponent
 
Pemetrexed disodium heptahydrate 97%
Catalog # APOSOR55746-5G
Supplier:  Apollo Scientific
CAS Number:  
Pemetrexed disodium heptahydrate 97%
Catalog # APOSOR55746-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :597.481 g/mol
Storage Temperature:Refrigerator
CAS Number:357166-29-1
Formula:C₂₀H₁₉N₅O₆·2Na·7H₂O
MDL Number:MFCD09029394