£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Northern Ireland
ContactUsLinkComponent
1,2-Bis(2,4,6-tribromophenoxy)ethane
1,2-Bis(2,4,6-tribromophenoxy)ethane 97%
1,2-Bis(2,4,6-tribromophenoxy)ethane 97%
APOSOR52482-1G
:
Apollo Scientific
:
1,2-Bis(2,4,6-tribromophenoxy)ethane 97%
APOSOR52482-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
687.636 g/mol
:
223,5…227 °C
:
253-692-3
:
GHS07
:
Ambient
:
37853-59-1
:
C₁₄H₈Br₆O₂
:
MFCD00071811