Methyl 2,4-dihydroxy-6-pentylbenzoate 95%
Catalog # APOSOR48127-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:238.28 g/mol
Storage Temperature:Ambient
CAS Number:58016-28-7
Formula:C₁₃H₁₈O₄
MDL Number:MFCD06737776