
3-Chloro-2-nitrophenol
Catalog # APOSOR400158-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:173.554 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:17802-02-7
Formula:C₆H₄ClNO₃
MDL Number:MFCD08703192