4,5,6,7-Tetrahydro-1H-indole
Catalog # APOSOR350389-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:121.18 g/mol
Storage Temperature:Ambient
CAS Number:13618-91-2
Formula:C₈H₁₁N
MDL Number:MFCD00012134