Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
 

2-(2-Bromophenyl)-thiazole 95+%

Supplier:   Apollo Scientific
2-(2-Bromophenyl)-thiazole 95+%
Supplier:  Apollo Scientific
Certificates
2-(2-Bromophenyl)-thiazole 95+%

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • CAS Number:30216-46-7
  • Formula:C₉H₆BrNS
  • MDL Number:MFCD19381944
  • Molecular Weight:240.12 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - £219.00
    £219.00
    1 * 1 g
       £219.00

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - £861.00
    £861.00
    1 * 5 g
       £861.00

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.