Order Entry
Northern Ireland
ContactUsLinkComponent
1-[4-(4-Chlorobenzoyl)piperidin-1-yl]ethan-1-one 95%
1-[4-(4-Chlorobenzoyl)piperidin-1-yl]ethan-1-one 95%
Catalog # APOSOR32361-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[4-(4-Chlorobenzoyl)piperidin-1-yl]ethan-1-one 95%
Catalog # APOSOR32361-1G
Supplier:  Apollo Scientific
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :265.735 g/mol
Storage Temperature:Ambient
CAS Number:59084-15-0
Formula:C₁₄H₁₆ClNO₂
MDL Number:MFCD01566224