Methyl 2-(4-iodophenoxy)acetate
Catalog # APOSOR310454-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:292.07 g/mol
Storage Temperature:Ambient
CAS Number:81720-18-5
Formula:C₉H₉IO₃
MDL Number:MFCD07113288