Order Entry
Northern Ireland
ContactUsLinkComponent
 
Irritant
1-(1-Ethoxyethyl)-1H-pyrazole
Catalog # APOSOR307169-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-(1-Ethoxyethyl)-1H-pyrazole
Catalog # APOSOR307169-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :140.183 g/mol
Flash Pt:>66 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:28791-95-9
Formula:C₇H₁₂N₂O
MDL Number:MFCD17676680