Order Entry
Northern Ireland
ContactUsLinkComponent
18-(Benzyloxy)-18-oxooctadecanoic acid
18-(Benzyloxy)-18-oxooctadecanoic acid
Catalog # APOSOR303206-500MG
Supplier:  Apollo Scientific
CAS Number:  
18-(Benzyloxy)-18-oxooctadecanoic acid
Catalog # APOSOR303206-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :404.583 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:189625-51-2
Formula:C₂₅H₄₀O₄
MDL Number:MFCD09991736