
N-(Chloroacetyl)glycine ethyl ester
Catalog # APOSOR28155-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:179.602 g/mol
Melting Pt:62…65 °C
:GHS07
Storage Temperature:Ambient
CAS Number:41602-50-0
Formula:C₆H₁₀ClNO₃
MDL Number:MFCD00216646