Order Entry
Northern Ireland
ContactUsLinkComponent
 
Corrosive
Irritant
3-[4-(Chloromethyl)phenyl]-1-methyl-1H-pyrazole 97%
Catalog # APOSOR12394-5G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
3-[4-(Chloromethyl)phenyl]-1-methyl-1H-pyrazole 97%
Catalog # APOSOR12394-5G
Supplier:  Apollo Scientific
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :206.671 g/mol
Melting Pt:78…80 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
 :GHS05
 :GHS07
CAS Number:916766-83-1
Formula:C₁₁H₁₁ClN₂
MDL Number:MFCD09702351