
4-(4-Bromophenyl)-2,4-dihydro-3H-1,2,4-triazol-3-one
Catalog # APOSOR110411-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:240.057 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:214117-50-7
Formula:C₈H₆BrN₃O
MDL Number:MFCD04125620