Order Entry
Northern Ireland
ContactUsLinkComponent
4-Isopropyl-2-nitroaniline
Irritant
4-Isopropyl-2-nitroaniline
Catalog # APOSOR0566-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Isopropyl-2-nitroaniline
Catalog # APOSOR0566-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :180.204 g/mol
Boiling Pt:141…143 °C (1 mm Hg)
Melting Pt:35…37 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:63649-64-9
Formula:C₉H₁₂N₂O₂
MDL Number:MFCD00092920