Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Terbium(III) oxalate decahydrate
Terbium(III) oxalate decahydrate 99,9%
Terbium(III) oxalate decahydrate 99,9%
APOSIN3484-10G
:
Apollo Scientific
:
Terbium(III) oxalate decahydrate 99,9%
APOSIN3484-10G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
10 g
Seq:
3236
:
:
762.06 g/mol (581.91 anhy)
:
Ambient
:
3288
:
6.1,III
:
GHS07
:
24670-06-2
:
3(C₂O₄)·Tb₂·10H₂O
:
MFCD00748140