

Vitamin-D3-[2H3] solution 1 mg/mL in ethanol
Catalog # APOSBISC1060A-1ML
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
Density:0,79 g/cm³ (25 °C)
Flash Pt:14 °C
ADR:3,II
:387.656 g/mol
UN:1170
:GHS02
:GHS07
Storage Temperature:Freezer
CAS Number:80666-48-4
Formula:C₂₇H₄₁D₃O
MDL Number:MFCD11656127