
Alternariol monomethyl ether 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:23452-05-3
:26894-49-5
Formula:C₁₅H₁₂O₅
MDL Number:MFCD00058532
:272.253 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together