ContactUsLinkComponent
PCR polishing kit
PCR polishing kit
Catalog # AGLS200409
Supplier:  AGILENT
CAS Number:  
PCR polishing kit
Catalog # AGLS200409
Supplier:  AGILENT
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Pk:
    1 kt
  • Description:
    PCR polishing kit
  • Details:
    Optimises blunt-end cloning efficiencies of PCR-generated fragments, generates blunt-ended DNA fragments in 30 minutes. Sufficient for 40 reactions.
Specifications
About this item
PCR polishing kit increases blunt-end cloning efficiencies associated with PCR-generated fragments.
  • Generates blunt-ended DNA fragments in 30 minutes
  • More efficient than Klenow polishing
  • Optimizes blunt-end cloning efficiencies
PCR Polishing Kit enables an increase in blunt-ended cloning efficiencies associated with polymerase chain reaction (PCR)-generated fragments. It polishes the ends of the 3’ overhang extensions of polymerase-generated DNA fragments directly from PCR amplification and can completely fill in 5’ overhangs to generate blunt ends.
Dramatic increases in the population of blunt-end DNA fragments following polishing with Agilent’s PCR Polishing Kit enables significant increase in overall experimental efficiency associated with procedures utilising blunt-ended ligation.