ABT-199 95%
Catalog # ACRO468231000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Yellow Powder or solid
- HPLC:≥94%
- MS:complies
- NMR (1H-NMR):Consistent with structure
Specification Test Results
About this item
:
:868.44 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:1257044-40-8
Formula:C₄₅H₅₀ClN₇O₇S
MDL Number:MFCD23160052