Styrene ≥99.5% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:100-42-5
:
Formula:C₈H₈
MDL Number:MFCD00008612
:104.15 g/mol
ADR:3,III
:Inhibited with tert-butylcatechol 10 - 15 ppm
UN:2055
:GHS02
:GHS07
Specification Test Results
- Assay (GC) :min 99,5%
- Identity (IR) :Passes test
- Appearance :clear liquid
- Density (d 20°C/4°C) :0,905 - 0,907
- Stabilised with 4-tert-Butylpyrocatechol:10 - 15 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC85999.180 spec_VWRC85999.290.pdf spec_VWRC85999.460
Product specification
Frequently Bought Together