Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Acid hematoxylin solution
Acid hematoxylin solution, Sigma-Aldrich®
Acid hematoxylin solution, Sigma-Aldrich®
Irritant
Acid hematoxylin solution, Sigma-Aldrich®
Catalog # 2852-100ML
Supplier:
SIGMA ALDRICH MICROSCOPY
CAS Number:
Expand image
Irritant
Acid hematoxylin solution, Sigma-Aldrich®
Catalog # 2852-100ML
Supplier:
SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
Pk:
100 mL
Seq:
2440
Specifications
About this item
Product Details
Documentation
:
:
GHS07
Storage Temperature:
Ambient
:
1
Product Details
Documentation