
Reactive Orange 16 ≥70 % (dye content), Sigma-Aldrich®
Catalog # 306509-100G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:617.54 g/mol
Melting Pt:>300 °C (lit.)
:GHS07
Storage Temperature:Ambient
CAS Number:12225-83-1
Formula:C₂H₁₇N₃Na₂O₁₁S₃
MDL Number:MFCD00013281