Azure B tetrafluoroborate 95% (dye content), Sigma-Aldrich®
Catalog # 302619-5G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:357.18 g/mol
Melting Pt:244 °C (dec., lit.)
Storage Temperature:Ambient
CAS Number:79288-94-1
Formula:C₁₅H₁₆BF₄N₃S
MDL Number:MFCD00011761