Order Entry
Northern Ireland
ContactUsLinkComponent
Sulphobromophthalein sodium salt hydrate, Sigma-Aldrich®
Health
Sulphobromophthalein sodium salt hydrate, Sigma-Aldrich®
Catalog # S0252-5G
CAS Number:  
undefined
Health
Sulphobromophthalein sodium salt hydrate, Sigma-Aldrich®
Catalog # S0252-5G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :837.98 g/mol (anhy)
 :GHS08
Storage Temperature:Ambient
CAS Number:71-67-0
Formula:C₂₀H₈Br₄O₁₀S₂Na₂·xH₂O
MDL Number:MFCD00150017