Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Carboxyfluorescein
6-Carboxyfluorescein, Sigma-Aldrich®
6-Carboxyfluorescein, Sigma-Aldrich®
C0662-50MG
:
SIGMA ALDRICH MICROSCOPY
:
6-Carboxyfluorescein, Sigma-Aldrich®
C0662-50MG
:
SIGMA ALDRICH MICROSCOPY
:
Pk:
50 mg
Seq:
2370
:
:
376.32 g/mol
:
>300 °C (lit.)
:
Refrigerator
:
3301-79-9
:
C₂₁H₁₂O₇
:
MFCD00036873