Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Trichrome stain
Trichrome stain solution LG, Sigma-Aldrich®
Trichrome stain solution LG, Sigma-Aldrich®
Trichrome stain solution LG, Sigma-Aldrich®
Catalog # HT10316-500ML
Supplier:
SIGMA ALDRICH MICROSCOPY
CAS Number:
Expand image
Trichrome stain solution LG, Sigma-Aldrich®
Catalog # HT10316-500ML
Supplier:
SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
Pk:
500 mL
Seq:
2725
Specifications
About this item
Product Details
Documentation
:
Storage Temperature:
Ambient
:
1
Product Details
Documentation