Order Entry
Northern Ireland
ContactUsLinkComponent
α-Naphthyl Acetate (Non-Specific Esterase) Kit Select reagents packaged in gelatin capsules., Sigma-Aldrich®
Flammable
Corrosive
Irritant
Health
α-Naphthyl Acetate (Non-Specific Esterase) Kit Select reagents packaged in gelatin capsules., Sigma-Aldrich®
Catalog # 90A1-1KT
CAS Number:  
Flammable
Corrosive
Irritant
Health
α-Naphthyl Acetate (Non-Specific Esterase) Kit Select reagents packaged in gelatin capsules., Sigma-Aldrich®
Catalog # 90A1-1KT
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Seq:
    0075

Specifications

About this item

 :
ADR:9,II
 :M-Clarity™ quality level = MQ500 (more info.)
Flash Pt:36 °C
UN:3316
 :GHS02
 :GHS05
 :GHS07
 :GHS08
Storage Temperature:Refrigerator
 :1
MDL Number:MFCD08705438