3-((2'-(1H-Tetrazol-5-yl)[1,1'-biphenyl]-4-yl)methyl)-2-butyl-1,3-diazaspiro[4.4]non-1-en-4-one
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:138402-11-6
:
Formula:C₂₅H₂₈N₆O
MDL Number:MFCD00864464
:428.5294 g/mol
Melting Pt:180…182 °C
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to almost white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:180 to 184 °C
- Loss on drying:≤2 %
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together