Aluminium oxide basic Brockmann Grade I 50-200 µm, 90A for chromatography
Catalog # ACRO463350010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Appearance:White to almost white Crystalline powder
- Particle size:≤10 % (< 50 um)
- Specific surface area:90 to 170 m2/g,N2 adsorp
- pH:8.5 to 10.5 (10 % suspension in water)
Specification Test Results
About this item
:
:101.96 g/mol
Boiling Pt:2977 °C (760 mmHg)
Melting Pt:2030 °C
Density:3,97 g/cm³
Storage Temperature:Ambient
CAS Number:1344-28-1
Formula:Al₂O₃
MDL Number:MFCD00003424