Order Entry
Northern Ireland
ContactUsLinkComponent
Lactophenol blue solution
Lactophenol blue solution
Catalog # RALA363060-0125_U
Supplier:  RAL
CAS Number:  
Lactophenol blue solution
Catalog # RALA363060-0125_U
Supplier:  RAL
CAS Number:  

Specifications

  • Pk:
    125 mL
  • Seq:
    2475

Specifications

About this item

 :
 :1
MDL Number:MFCD00146574