Order Entry
Northern Ireland
ContactUsLinkComponent
 
Health
Irritant
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), gauze woven from wire 100 mesh Ø 0.11 mm (0.0045 in) Type 316
Catalog # 46703.HB
CAS Number:  
Health
Irritant
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), gauze woven from wire 100 mesh Ø 0.11 mm (0.0045 in) Type 316
Catalog # 46703.HB
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    1886
  • Pk Info:
    300×300 mm

Specifications

About this item

 :
 :55.845 g/mol
 :GHS08
 :GHS07
Storage Temperature:Ambient
Formula:Fe
MDL Number:MFCD00148499