Order Entry
Northern Ireland
ContactUsLinkComponent
Sodium sulphate, anhydrous ≥99.0%, powder, AR® ACS, Macron Fine Chemicals™
Sodium sulphate, anhydrous ≥99.0%, powder, AR® ACS, Macron Fine Chemicals™
Catalog # MACR8020-06
CAS Number:  
undefined
Sodium sulphate, anhydrous ≥99.0%, powder, AR® ACS, Macron Fine Chemicals™
Catalog # MACR8020-06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber
  • Pk:
    2,5 kg
  • Seq:
    4076
  • Pk Info:
    1×2,5 kg

Specifications

Specification Test Results

  • ACS - Assay (Na₂SO₄):
    ≥99.0 %
  • ACS - Appearance (white, crystalline powder):
    Passes Test
  • Solubility and Foreign Matter:
    Passes Test
  • ACS - Calcium (Ca):
    ≤0.01 %
  • ACS - Chloride (Cl):
    ≤0.001 %
  • ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • ACS - Insoluble Matter:
    ≤0.01 %
  • ACS - Iron (Fe):
    ≤0.001 %
  • ACS - Loss on Ignition:
    ≤0.5 %
  • ACS - Magnesium (Mg):
    ≤0.005 %
  • ACS - Nitrogen Compounds (as N):
    ≤5 ppm
  • ACS - pH of 5% Solution at 25°C:
    5.2 - 9.2
  • ACS - Phosphate (PO₄):
    ≤0.001 %
  • ACS - Potassium (K):
    ≤0.002 %
  • Thru U.S. No. 50 Sieve:
    ≥99.0 %

Specification Test Results

About this item

 :
Merck Index:12,08829
Boiling Pt:1429 °C (1013 hPa)
Melting Pt:888 °C
Density:2,7 g/cm³ (20 °C)
 :Management of Change (MOC) category = N
 :142.04 g/mol
Storage Temperature:Ambient
EINECS:231-820-9
CAS Number:7757-82-6
Formula:Na₂SO₄
MDL Number:MFCD00003504