
Sodium hydroxide 97.0-100.5% alkali metal, pellets, GenAR® NF, FCC, Macron Fine Chemicals™
:1310-73-2
:
:NaOH
:MFCD00003548
:40 g/mol
:8,II
:Management of Change (MOC) category = R
:1823
:GHS05
- NF - Total Alkali:95.0 - 100.5 %
- NF - Content of Sodium:54.0 - 59.8 %
- NF - Insoluble Substances and Organic Matter:Passes Test
- NF - Identification A (Sodium):Passes Test
- NF - Identification B (pH):≥11.0
- NF - Potassium (K):≤0.5 %
- NF/FCC - Carbonate (as Na₂CO₃):≤3.0 %
- BP/EP - Assay (Total alkali as NaOH):97.0 - 100.5 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Iron (Fe):≤10 ppm
- EP/BP - Carbonate (as Na₂CO₃):≤2.0 %
- EP/BP - Chloride (Cl):≤200 ppm
- EP/BP - Sulfate (SO₄):≤200 ppm
Frequently Bought Together