Phenylhydrazinium chloride ≥99%, AnalaR NORMAPUR® analytical reagent
Catalog # 87096.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:≥ 99 %
- White to faint beige powder:Passes test
- Solubility in water:Passes test
- Loss on drying (105°C):≤ 0.5 %
- Ignition residue (SO4):≤ 0.1 %
- SO4 (Sulphate):≤ 100 ppm
- Fe (Iron):≤ 10 ppm
Specification Test Results
About this item
Synonyms: Phenylhydrazine hydrochloride, Phenylhydrazinium chloride
:
:144.6 g/mol
EINECS:200-444-7
UN:2811
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
CAS Number:59-88-1
Formula:C₆H₉ClN₂
MDL Number:MFCD00012924
Melting Pt:243…246 °C
ADR:6.1 III
Merck Index:12,07447