Accessories for ion selective electrodes, Orion™
Accessories for solid state and plastic membrane ion selective electrodes.
- Electrodes handles are used with plastic membrane combination or half-cell electrodes
- Polishing strips help restore the performance of solid-state, half-cell, or combination ion selective electrodes
Frequently Bought Together