Order Entry
Northern Ireland
ContactUsLinkComponent
Analytical standard, Trifloxystrobin, reference material, Fluka™
Analytical standard, Trifloxystrobin, reference material, Fluka™
Catalog # HONCR1765-100MG
CAS Number:  
Analytical standard, Trifloxystrobin, reference material, Fluka™
Catalog # HONCR1765-100MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • Standards:
    Reference material
  • CAS No.:
    141517-21-7
  • Description:
    Trifloxystrobin
  • Description:
    Trifloxystrobin
  • Form:
    Neat

Specifications

About this item