Analytical standard, Propetamphos, reference material, Fluka™
Catalog # HONCR1713-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Standards:Reference material
- CAS No.:31218-83-4
- Description:Propetamphos
- Storage temperature:2…8 °C
- Description:Propetamphos
- Form:Neat