Analytical standard, Primisulfuron-methyl, reference material, Fluka™
Catalog # HONCR1711-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Standards:Reference material
- CAS No.:86209-51-0
- Description:Primisulfuron-methyl
- Description:Primisulfuron-methyl
- Form:Neat