Order Entry
Northern Ireland
ContactUsLinkComponent
Analytical standard, alpha-Cypermethrin, reference material, Fluka™
Analytical standard, alpha-Cypermethrin, reference material, Fluka™
Catalog # HONCR1787-100MG
CAS Number:  
Analytical standard, alpha-Cypermethrin, reference material, Fluka™
Catalog # HONCR1787-100MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • Standards:
    Reference material
  • CAS No.:
    67375-30-8
  • Description:
    alpha-Cypermethrin
  • Description:
    alpha-Cypermethrin
  • Form:
    Neat

Specifications

About this item