Analytical standard, Fludioxonil, reference material, Fluka™
Catalog # HONCR1784-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Standards:Reference material
- CAS No.:131341-86-1
- Description:Fludioxonil
- Description:Fludioxonil
- Form:Neat