Analytical standard, Fenpropathrin, reference material, Fluka™
Catalog # HONCR1776-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Standards:Reference material
- CAS No.:39515-41-8
- Description:Fenpropathrin
- Formula:C₁₂H₁₂Br₂N₂ ·H₂O
- Description:Fenpropathrin
- Form:Neat