



n-Butyllithium 2.5M in hexane
Catalog # H36949.AE
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, septum-sealed
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:64.06 g/mol
ADR:4.2,I
:Packaged under Nitrogen in resealable AcroSeal® bottles
Flash Pt:–21°C (–6°F)
EINECS:203-698-7
UN:3394
:GHS02
:GHS05
:GHS08
:GHS09
Storage Temperature:Ambient
CAS Number:109-72-8
Formula:C₄H₉Li
MDL Number:MFCD00009414