Diethylzinc ∼15% (w/w) in hexane
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:557-20-0
:
Formula:C₄H₁₀Zn
MDL Number:MFCD00009021
:123.51 g/mol
ADR:4.2,I
:Packaged under Nitrogen in resealable AcroSeal® bottles
Flash Pt:–40°C (–40°F)
EINECS:209-161-3
UN:3394
:GHS02
:GHS05
:GHS08
:GHS09
Storage Temperature:Ambient
Frequently Bought Together